Product Details

SNP ID
rs11866302
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2967853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAAGGGCCCCCGGCGCTGGGG[C/G]CTTCCAGGGGCCCCAGGAGAAGCTG
Phenotype
MIM: 609899 MIM: 614578 MIM: 602474
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
KREMEN2 PubMed Links
Additional Information
For this assay, SNP(s) [rs117373306] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KREMEN2
Gene Name
kringle containing transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253725.1 1527 Silent Mutation GGC,GGG G342G NP_001240654.1
NM_001253726.1 1527 Missense Mutation CCT,GCT P369A NP_001240655.1
NM_024507.3 1527 Silent Mutation GGC,GGG G381G NP_078783.1
NM_172229.2 1527 Missense Mutation CCT,GCT P408A NP_757384.1
Gene
PAQR4
Gene Name
progestin and adipoQ receptor family member 4
There are no transcripts associated with this gene.

Gene
PKMYT1
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
There are no transcripts associated with this gene.

View Full Product Details