Product Details

SNP ID
rs4807219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2275472 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGAGGCGCTGGCCAGCCCCCAG[C/T]CCTGGAGGGGCAGCTGCATTCCTGG
Phenotype
MIM: 601579
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf35 PubMed Links

Gene Details

Gene
C19orf35
Gene Name
chromosome 19 open reading frame 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198532.2 1730 UTR 3 NP_940934.1
Gene
OAZ1
Gene Name
ornithine decarboxylase antizyme 1
There are no transcripts associated with this gene.

View Full Product Details