Product Details

SNP ID
rs2023885
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:48049011 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGAGAACGGGAGGGTAGCGTAAGCA[A/G]GAATTCTGGGTCGCAATTGTGTTTA
Phenotype
MIM: 615526 MIM: 163260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPZ2 PubMed Links

Gene Details

Gene
COPZ2
Gene Name
coatomer protein complex subunit zeta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016429.3 Intron NP_057513.1
XM_011524874.2 Intron XP_011523176.1
Gene
NFE2L1
Gene Name
nuclear factor, erythroid 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003204.2 Intron NP_003195.1
XM_005257410.3 Intron XP_005257467.1
XM_005257411.3 Intron XP_005257468.1
XM_005257412.3 Intron XP_005257469.1
XM_005257413.3 Intron XP_005257470.1
XM_005257415.3 Intron XP_005257472.1
XM_017024691.1 Intron XP_016880180.1

View Full Product Details