Product Details

SNP ID
rs2040786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26169116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGCAAATTATATTACAAATTCC[A/G]TAGGCCTTTTTTACTTTGTAAATAT
Phenotype
MIM: 604135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFE2L3 PubMed Links

Gene Details

Gene
NFE2L3
Gene Name
nuclear factor, erythroid 2 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004289.6 Intron NP_004280.5

View Full Product Details