Product Details

SNP ID
rs6465762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100022931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAAACACCCTGAGGGTGGAGTC[A/G]GCCTGTTTGAGGTTTCTGTTTGCAA
Phenotype
MIM: 601260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN1 PubMed Links

Gene Details

Gene
ZKSCAN1
Gene Name
zinc finger with KRAB and SCAN domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287054.1 Intron NP_001273983.1
NM_001287055.1 Intron NP_001273984.1
NM_003439.2 Intron NP_003430.1
XM_005250565.4 Intron XP_005250622.1
XM_011516559.2 Intron XP_011514861.1

View Full Product Details