Product Details
- SNP ID
-
rs2074574
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:985997 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAGGTAAGAGGAGCAAAGCGTGAG[C/G]GTTTCCCACAGGACATCTCAGCCCA
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
WDR18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80201716] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WDR18
- Gene Name
- WD repeat domain 18
View Full Product Details