Product Details
- SNP ID
-
rs10405599
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:986825 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGAGTTTGGCCCATACCGTAGCTG[C/T]GGCGTTAGATCACCGGCTGTGTTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR18
PubMed Links
Gene Details
- Gene
- WDR18
- Gene Name
- WD repeat domain 18
View Full Product Details