Product Details
- SNP ID
-
rs3094835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56981942 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTAACTTCTTACCCCCAGAAAA[C/G]CTGATCTTTTGAAGGGACTGCTACA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SCPEP1
PubMed Links
Gene Details
- Gene
- SCPEP1
- Gene Name
- serine carboxypeptidase 1
View Full Product Details