Product Details

SNP ID
rs1548624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98889676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCCTTTTACTTCAGCCCTCTGAG[C/T]AGCTAAGACCACAGCATAGGCACGT
Phenotype
MIM: 603015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3609 PubMed Links

Gene Details

Gene
MIR3609
Gene Name
microRNA 3609
There are no transcripts associated with this gene.

Gene
SCARNA28
Gene Name
small Cajal body-specific RNA 28
There are no transcripts associated with this gene.

Gene
TRRAP
Gene Name
transformation/transcription domain associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244580.1 Intron NP_001231509.1
NM_003496.3 Intron NP_003487.1

View Full Product Details