Product Details
- SNP ID
-
rs2072227
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:17494491 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCACTCTGGCAGCACAAGGCCCTG[C/T]CACCCTTTGGAGACCCCTCTGGGTG
- Phenotype
-
MIM: 605242
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
USH1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113336938] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- USH1C
- Gene Name
- USH1 protein network component harmonin
View Full Product Details