Product Details

SNP ID
rs174545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61801834 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTTTGCAAATGAGGATGGAGAAG[C/G]CACTGAGAAATGGACCAGTGAGCTG
Phenotype
MIM: 606148 MIM: 600393
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FADS1 PubMed Links

Gene Details

Gene
FADS1
Gene Name
fatty acid desaturase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013402.4 1885 UTR 3 NP_037534.3
XM_011545022.2 1885 UTR 3 XP_011543324.1
Gene
FEN1
Gene Name
flap structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
MIR611
Gene Name
microRNA 611
There are no transcripts associated with this gene.

Gene
TMEM258
Gene Name
transmembrane protein 258
There are no transcripts associated with this gene.

View Full Product Details