Product Details
- SNP ID
-
rs35232737
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18557829 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGCTCCAGGGTACGTCCCCGCCA[C/G]GCGCGCTCGCAGGATCGGTGCGTGG
- Phenotype
-
MIM: 615178
MIM: 191321
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KXD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115804630] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KXD1
- Gene Name
- KxDL motif containing 1
- Gene
- UBA52
- Gene Name
- ubiquitin A-52 residue ribosomal protein fusion product 1
There are no transcripts associated with this gene.
View Full Product Details