Product Details

SNP ID
rs9935429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2160406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACTCCTGGAGCATCCAGCATGG[A/G]CAGTGTGGACGGGCAGGTGGTGTGG
Phenotype
MIM: 605455 MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB26 PubMed Links

Gene Details

Gene
RAB26
Gene Name
RAB26, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
SNHG19
Gene Name
small nucleolar RNA host gene 19
There are no transcripts associated with this gene.

Gene
SNORD60
Gene Name
small nucleolar RNA, C/D box 60
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032271.2 Intron NP_115647.2
XM_005255627.4 Intron XP_005255684.1
XM_011522700.1 Intron XP_011521002.1

View Full Product Details