Product Details

SNP ID
rs12939333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9898413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCATGACCCTTGGAGCTCCCTG[C/G]AGACCAGCAAAGAGCGGTGAATGAC
Phenotype
MIM: 603659 MIM: 179618
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLP2R PubMed Links

Gene Details

Gene
GLP2R
Gene Name
glucagon like peptide 2 receptor
There are no transcripts associated with this gene.

Gene
RCVRN
Gene Name
recoverin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002903.2 Intron NP_002894.1

View Full Product Details