Product Details
- SNP ID
-
rs34877480
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39682838 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTGTAGCAGGGTTACCCAAAAT[A/T]ATTGAACTTGATCCTTTTGATGGAC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS17A
PubMed Links
Gene Details
- Gene
- LGALS17A
- Gene Name
- galectin 14 pseudogene
There are no transcripts associated with this gene.
View Full Product Details