Product Details

SNP ID
rs35192500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34271131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCCTCAATGGACTTTTCTTCTT[C/G]TTGTTTCATTGCAGTTGGGATTAAT
Phenotype
MIM: 158106
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCL7 PubMed Links

Gene Details

Gene
CCL7
Gene Name
C-C motif chemokine ligand 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006273.3 Intron NP_006264.2

View Full Product Details