Product Details
- SNP ID
-
rs17709621
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39784545 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGCAGAAGGGCCAAGGCGTTATC[A/G]TCGGCCACGCACAAGATTTCTCTCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LEUTX
PubMed Links
Gene Details
- Gene
- LEUTX
- Gene Name
- leucine twenty homeobox
View Full Product Details