Product Details

SNP ID
rs8050273
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:27569768 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACTGGGCCAACCAGTCTTCTAGGG[A/C]GTTTAGATAATAAATGCTCTTAATT
Phenotype
MIM: 616650
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0556 PubMed Links

Gene Details

Gene
KIAA0556
Gene Name
KIAA0556
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015202.3 Intron NP_056017.3
XM_005255201.3 Intron XP_005255258.1
XM_005255202.3 Intron XP_005255259.1
XM_005255203.3 Intron XP_005255260.1
XM_005255206.3 Intron XP_005255263.1
XM_005255208.2 Intron XP_005255265.1
XM_006721025.3 Intron XP_006721088.1
XM_011545773.2 Intron XP_011544075.1
XM_011545774.1 Intron XP_011544076.1
XM_011545775.2 Intron XP_011544077.1
XM_011545776.2 Intron XP_011544078.1
XM_011545777.2 Intron XP_011544079.1
XM_017023085.1 Intron XP_016878574.1
XM_017023086.1 Intron XP_016878575.1
XM_017023087.1 Intron XP_016878576.1
XM_017023088.1 Intron XP_016878577.1

View Full Product Details