Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040113.1 | 6467 | UTR 3 | NP_001035202.1 | ||
NM_001040114.1 | 6467 | UTR 3 | NP_001035203.1 | ||
NM_002474.2 | 6467 | UTR 3 | NP_002465.1 | ||
NM_022844.2 | 6467 | UTR 3 | NP_074035.1 | ||
XM_011522502.2 | 6467 | Intron | XP_011520804.1 | ||
XM_017023250.1 | 6467 | Intron | XP_016878739.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001143979.1 | 6467 | Intron | NP_001137451.1 | ||
NM_017668.2 | 6467 | Intron | NP_060138.1 | ||
XM_005255396.4 | 6467 | Intron | XP_005255453.1 | ||
XM_006720897.3 | 6467 | Intron | XP_006720960.1 | ||
XM_006720900.3 | 6467 | Intron | XP_006720963.1 | ||
XM_011522553.2 | 6467 | Intron | XP_011520855.1 | ||
XM_017023349.1 | 6467 | Intron | XP_016878838.1 | ||
XM_017023350.1 | 6467 | Intron | XP_016878839.1 | ||
XM_017023351.1 | 6467 | Intron | XP_016878840.1 | ||
XM_017023352.1 | 6467 | Intron | XP_016878841.1 | ||
XM_017023353.1 | 6467 | Intron | XP_016878842.1 | ||
XM_017023354.1 | 6467 | Intron | XP_016878843.1 | ||
XM_017023355.1 | 6467 | Intron | XP_016878844.1 | ||
XM_017023356.1 | 6467 | Intron | XP_016878845.1 | ||
XM_017023357.1 | 6467 | Intron | XP_016878846.1 |