Product Details

SNP ID
rs13055536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23078727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGCACAGGAAGAAGGCCAAGGG[G/T]GAGGGCCAGGGCTTCTAAGGGGGCT
Phenotype
MIM: 139160 MIM: 605663
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GNAZ PubMed Links

Gene Details

Gene
GNAZ
Gene Name
G protein subunit alpha z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002073.2 Intron NP_002064.1
XM_017028786.1 Intron XP_016884275.1
Gene
RSPH14
Gene Name
radial spoke head 14 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014433.2 Intron NP_055248.1
XM_011530149.2 Intron XP_011528451.1
XM_011530150.1 Intron XP_011528452.1
XM_011530151.2 Intron XP_011528453.1
XM_011530152.2 Intron XP_011528454.1
XM_011530154.2 Intron XP_011528456.1
XM_011530155.2 Intron XP_011528457.1
XM_017028774.1 Intron XP_016884263.1

View Full Product Details