Product Details

SNP ID
rs1010167
Assay Type
DME
NCBI dbSNP Submissions
46
Location
Chr.1:109656105 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
TGGCGAGGCCGAGCCCCTCCTAGTG[C/G]TTCCGGACCTTGCTCCCTGAACACT
Phenotype
MIM: 138333
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GSTM4 PubMed Links

Gene Details

Gene
GSTM4
Gene Name
glutathione S-transferase mu 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000850.4 30 UTR 5 NP_000841.1
NM_147148.2 30 UTR 5 NP_671489.1
XM_017001085.1 30 UTR 5 XP_016856574.1
XM_017001086.1 30 UTR 5 XP_016856575.1
XM_017001087.1 30 UTR 5 XP_016856576.1
XM_017001088.1 30 UTR 5 XP_016856577.1

View Full Product Details