Product Details

SNP ID
rs12158466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29768522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTGCCCCATATGAAAGCATTGC[C/T]ACTGTCCCATCTCTTGCAAGTGGTT
Phenotype
MIM: 610843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UQCR10 PubMed Links

Gene Details

Gene
UQCR10
Gene Name
ubiquinol-cytochrome c reductase, complex III subunit X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003684.1 Intron NP_001003684.1
NM_013387.3 Intron NP_037519.2
Gene
ZMAT5
Gene Name
zinc finger matrin-type 5
There are no transcripts associated with this gene.

View Full Product Details