Product Details

SNP ID
rs982085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138439892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACCCCGCTGGGGGAGGACGCTC[C/G]GTGGCCTTCATTCCAGGCTGCCAGC
Phenotype
MIM: 609373 MIM: 609347
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KDM3B PubMed Links

Gene Details

Gene
KDM3B
Gene Name
lysine demethylase 3B
There are no transcripts associated with this gene.

Gene
REEP2
Gene Name
receptor accessory protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271803.1 Intron NP_001258732.1
NM_016606.3 Intron NP_057690.2

View Full Product Details