Product Details
- SNP ID
-
rs5997898
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31176977 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATCATGGAGCCCTTTGGAAGGTT[A/C]GGCAGATCTGGGTTAGGTACCCTAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNF185
PubMed Links
Gene Details
- Gene
- RNF185
- Gene Name
- ring finger protein 185
View Full Product Details