Product Details
- SNP ID
-
rs4764541
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6204245 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACACCCCTACACATACCACATACA[A/G]CAATAGAACTCTCCTACACAACCCA
- Phenotype
-
MIM: 143030
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CD9
PubMed Links
Gene Details
- Gene
- CD9
- Gene Name
- CD9 molecule
View Full Product Details