Product Details

SNP ID
rs7713839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137874962 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCTGGTAAATGTCTAACAATCA[C/T]TGGACAAAAGAGCCATTATTTGTGA
Phenotype
MIM: 604103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYOT PubMed Links

Gene Details

Gene
MYOT
Gene Name
myotilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135940.1 Intron NP_001129412.1
NM_001300911.1 Intron NP_001287840.1
NM_006790.2 Intron NP_006781.1
XM_017010060.1 Intron XP_016865549.1
XM_017010061.1 Intron XP_016865550.1
XM_017010062.1 Intron XP_016865551.1

View Full Product Details