Product Details

SNP ID
rs67616912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56843609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGAAGAGCCAACATTATCTTTT[G/T]TCTTAGAGGACAATTAAATATAGGT
Phenotype
MIM: 601483
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIMT1 PubMed Links

Gene Details

Gene
MIMT1
Gene Name
MER1 repeat containing imprinted transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
PEG3
Gene Name
paternally expressed 3
There are no transcripts associated with this gene.

Gene
ZIM2
Gene Name
zinc finger imprinted 2
There are no transcripts associated with this gene.

View Full Product Details