Product Details

SNP ID
rs1005170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3713178 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTGAGCCGGCCACACCTAGAAT[C/G]TGCCCTTCCTCAAAGGTCGTTTGGA
Phenotype
MIM: 612689
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TJP3 PubMed Links

Gene Details

Gene
TJP3
Gene Name
tight junction protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267560.1 Intron NP_001254489.1
NM_001267561.1 Intron NP_001254490.1

View Full Product Details