Product Details

SNP ID
rs11980923
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90407559 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGCAAATAATCGAGTGCCTTTTA[C/T]GTGCCAGTATAGGGAACAGTGAACA
Phenotype
MIM: 611232
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CLDN12 PubMed Links
Additional Information
For this assay, SNP(s) [rs139136039] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CLDN12
Gene Name
claudin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185072.2 Intron NP_001172001.1
NM_001185073.2 Intron NP_001172002.1
NM_012129.4 Intron NP_036261.1
Gene
LOC101409256
Gene Name
cell division cycle 42 pseudogene
There are no transcripts associated with this gene.

Gene
LOC102723899
Gene Name
pituitary tumor-transforming gene 1 protein-interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011516764.2 Intron XP_011515066.1
XM_017012889.1 Intron XP_016868378.1
XM_017012890.1 Intron XP_016868379.1
XM_017012891.1 Intron XP_016868380.1
XM_017012892.1 Intron XP_016868381.1
XM_017012893.1 Intron XP_016868382.1
XM_017012894.1 Intron XP_016868383.1

View Full Product Details