Product Details

SNP ID
rs10770810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21441901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGAAAGACCTTCCCCTATGGGGA[G/T]GGCTGCTTGCTAGATGGGATGTAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PYROXD1 PubMed Links

Gene Details

Gene
PYROXD1
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024854.3 Intron NP_079130.2
XM_006719153.3 Intron XP_006719216.1
XM_017019976.1 Intron XP_016875465.1

View Full Product Details