Product Details

SNP ID
rs2241042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17086809 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCCAGGTGGTGGAAAGGGCCTG[A/C]GCCAAGGTCCTTAAGTGGGACAGGG
Phenotype
MIM: 605461
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL17RA PubMed Links

Gene Details

Gene
IL17RA
Gene Name
interleukin 17 receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289905.1 Intron NP_001276834.1
NM_014339.6 Intron NP_055154.3

View Full Product Details