Product Details

SNP ID
rs2110581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46353103 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCAAGCCTGGCTCTGCAGGTGCA[A/G]ACAGGGCATTTGACCTTGAGCAAGA
Phenotype
MIM: 609976 MIM: 600658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIF3A PubMed Links

Gene Details

Gene
HIF3A
Gene Name
hypoxia inducible factor 3 alpha subunit
There are no transcripts associated with this gene.

Gene
PPP5C
Gene Name
protein phosphatase 5 catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204284.1 Intron NP_001191213.1
NM_006247.3 Intron NP_006238.1
XM_017026935.1 Intron XP_016882424.1

View Full Product Details