Product Details

SNP ID
rs10777822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96914785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTTGCCTTGCACCTTGAGCAGT[A/T]CAGGATTCCTGTAATAACCACACAA
Phenotype
MIM: 600372
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NEDD1 PubMed Links

Gene Details

Gene
NEDD1
Gene Name
neural precursor cell expressed, developmentally down-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135175.1 Intron NP_001128647.1
NM_001135176.1 Intron NP_001128648.1
NM_001135177.1 Intron NP_001128649.1
NM_152905.3 Intron NP_690869.1
XM_005268644.2 Intron XP_005268701.1
XM_006719237.3 Intron XP_006719300.1
XM_011537903.2 Intron XP_011536205.1
XM_011537904.2 Intron XP_011536206.1
XM_011537905.2 Intron XP_011536207.1
XM_011537906.2 Intron XP_011536208.1
XM_017018801.1 Intron XP_016874290.1
XM_017018802.1 Intron XP_016874291.1

View Full Product Details