Product Details

SNP ID
rs12457540
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:51036434 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCAGTCCTTGTAAAAATTGAATAAT[C/G]TAGTGAAAGGATGACCGGAAATAAT
Phenotype
MIM: 600993
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SMAD4 PubMed Links
Additional Information
For this assay, SNP(s) [rs78801230] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SMAD4
Gene Name
SMAD family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005359.5 Intron NP_005350.1

View Full Product Details