Product Details
- SNP ID
-
rs34038511
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:79619302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTTATCTAAACCTAGTCCCACAA[A/G]TACTTTTACAAGAGAGCATGTTAAA
- Phenotype
-
MIM: 600621
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STMN2
PubMed Links
Gene Details
- Gene
- STMN2
- Gene Name
- stathmin 2
View Full Product Details