Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001178074.1 | Intron | NP_001171545.1 | |||
NM_021150.3 | Intron | NP_066973.2 | |||
XM_005268754.4 | Intron | XP_005268811.1 | |||
XM_005268757.4 | Intron | XP_005268814.1 | |||
XM_011538093.2 | Intron | XP_011536395.1 | |||
XM_017019098.1 | Intron | XP_016874587.1 | |||
XM_017019099.1 | Intron | XP_016874588.1 | |||
XM_017019100.1 | Intron | XP_016874589.1 | |||
XM_017019101.1 | Intron | XP_016874590.1 | |||
XM_017019102.1 | Intron | XP_016874591.1 |