Product Details

SNP ID
rs1151763
Assay Type
Validated
NCBI dbSNP Submissions
44
Location
Chr.1:61691119 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCACATATCCTCATCTTTCTGATT[C/T]GTGGCTGTTTGTCTCCACTTAATAT
Phenotype
MIM: 610080
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TM2D1 PubMed Links
Additional Information
For this assay, SNP(s) [rs35138123] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TM2D1
Gene Name
TM2 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032027.2 Intron NP_114416.1

View Full Product Details