Product Details

SNP ID
rs11970677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41908544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCACATTTTTGCTTCCCAACCT[A/G]TCTTAAAGCATACTAAGGAAAGGTA
Phenotype
MIM: 612915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED20 PubMed Links

Gene Details

Gene
MED20
Gene Name
mediator complex subunit 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305455.1 Intron NP_001292384.1
NM_001305456.1 Intron NP_001292385.1
NM_001305457.1 Intron NP_001292386.1
NM_004275.4 Intron NP_004266.2

View Full Product Details