Product Details

SNP ID
rs2454217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:126917654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGTTATGAAGAATAAATGAGAC[A/G]GGAGGGGAGAAGGCACCTGATCCAC
Phenotype
MIM: 614444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RALGPS1 PubMed Links

Gene Details

Gene
RALGPS1
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190728.1 Intron NP_001177657.1
NM_001190729.1 Intron NP_001177658.1
NM_001190730.1 Intron NP_001177659.1
NM_001322320.1 Intron NP_001309249.1
NM_001322321.1 Intron NP_001309250.1
NM_001322322.1 Intron NP_001309251.1
NM_001322323.1 Intron NP_001309252.1
NM_001322324.1 Intron NP_001309253.1
NM_001322325.1 Intron NP_001309254.1
NM_014636.2 Intron NP_055451.1
XM_006717328.3 Intron XP_006717391.2
XM_011519225.1 Intron XP_011517527.1
XM_011519226.1 Intron XP_011517528.1
XM_011519227.1 Intron XP_011517529.1
XM_011519228.1 Intron XP_011517530.1
XM_011519229.1 Intron XP_011517531.1
XM_011519230.2 Intron XP_011517532.1
XM_011519231.1 Intron XP_011517533.1
XM_011519232.1 Intron XP_011517534.1
XM_011519233.1 Intron XP_011517535.1
XM_011519234.2 Intron XP_011517536.1
XM_011519235.2 Intron XP_011517537.1
XM_011519236.1 Intron XP_011517538.1
XM_011519238.2 Intron XP_011517540.1
XM_017015342.1 Intron XP_016870831.1
XM_017015343.1 Intron XP_016870832.1
XM_017015344.1 Intron XP_016870833.1
XM_017015345.1 Intron XP_016870834.1
XM_017015346.1 Intron XP_016870835.1
XM_017015347.1 Intron XP_016870836.1
XM_017015348.1 Intron XP_016870837.1
XM_017015349.1 Intron XP_016870838.1
XM_017015350.1 Intron XP_016870839.1

View Full Product Details