Product Details

SNP ID
rs9562321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41570717 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGCATGCATTGTCTGGAGGTAA[A/G]AGAAGTTGCACAAAAGCCATGGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR5006 PubMed Links

Gene Details

Gene
MIR5006
Gene Name
microRNA 5006
There are no transcripts associated with this gene.

Gene
VWA8
Gene Name
von Willebrand factor A domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009814.1 Intron NP_001009814.1
NM_015058.1 Intron NP_055873.1
XM_006719791.3 Intron XP_006719854.1
XM_011535007.2 Intron XP_011533309.1
XM_017020469.1 Intron XP_016875958.1
XM_017020470.1 Intron XP_016875959.1
XM_017020471.1 Intron XP_016875960.1
XM_017020472.1 Intron XP_016875961.1
XM_017020473.1 Intron XP_016875962.1
XM_017020474.1 Intron XP_016875963.1

View Full Product Details