Product Details
- SNP ID
-
rs9507040
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:23166769 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGTCATCACAGAAAAACCTTTTT[C/T]ATTTGTAAGGTTTGTAGGTACATAG
- Phenotype
-
MIM: 608896
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SGCG
PubMed Links
Gene Details
- Gene
- SGCG
- Gene Name
- sarcoglycan gamma
View Full Product Details