Product Details

SNP ID
rs10820329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96316009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTGAAACCCATAATGCAATATG[A/C]AAATATCTTTAATTCTATTGGTAAC
Phenotype
MIM: 605573 MIM: 609182
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSD17B3 PubMed Links

Gene Details

Gene
HSD17B3
Gene Name
hydroxysteroid 17-beta dehydrogenase 3
There are no transcripts associated with this gene.

Gene
SLC35D2
Gene Name
solute carrier family 35 member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286990.1 Intron NP_001273919.1
NM_007001.2 Intron NP_008932.2
XM_005251673.1 Intron XP_005251730.1
XM_005251674.1 Intron XP_005251731.1
XM_005251675.2 Intron XP_005251732.1
XM_005251676.1 Intron XP_005251733.1
XM_005251678.4 Intron XP_005251735.1
XM_006716939.2 Intron XP_006717002.1
XM_011518164.1 Intron XP_011516466.1

View Full Product Details