Product Details

SNP ID
rs11580710
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:145945371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCCTCTTTCCAAAAATAACAGA[C/A]ATCAAATTACTATAATGTTTCAGTT
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
LIX1L PubMed Links

Gene Details

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153713.2 Intron NP_714924.1
XM_017000316.1 Intron XP_016855805.1

View Full Product Details