Product Details
- SNP ID
-
rs756186
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:130399802 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGAGGGCACAAGGCCTTTCTGCCC[A/G]TTCAGCTCCCCCTAAAACACCAGGG
- Phenotype
-
MIM: 605571
MIM: 611602
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIWIL1
PubMed Links
Gene Details
- Gene
- PIWIL1
- Gene Name
- piwi like RNA-mediated gene silencing 1
- Gene
- RIMBP2
- Gene Name
- RIMS binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015347.4 |
4466 |
Silent Mutation |
AAC,AAT |
N1003N |
NP_056162.4 |
XM_011538102.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1259N |
XP_011536404.1 |
XM_011538103.2 |
4466 |
Silent Mutation |
AAC,AAT |
N1380N |
XP_011536405.1 |
XM_011538104.2 |
4466 |
Silent Mutation |
AAC,AAT |
N1259N |
XP_011536406.1 |
XM_011538105.2 |
4466 |
Silent Mutation |
AAC,AAT |
N1141N |
XP_011536407.1 |
XM_011538106.2 |
4466 |
Silent Mutation |
AAC,AAT |
N1088N |
XP_011536408.1 |
XM_011538107.2 |
4466 |
Intron |
|
|
XP_011536409.1 |
XM_011538108.2 |
4466 |
Intron |
|
|
XP_011536410.1 |
XM_017019105.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1259N |
XP_016874594.1 |
XM_017019106.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1141N |
XP_016874595.1 |
XM_017019107.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1020N |
XP_016874596.1 |
XM_017019108.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1020N |
XP_016874597.1 |
XM_017019109.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1020N |
XP_016874598.1 |
XM_017019110.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1020N |
XP_016874599.1 |
XM_017019111.1 |
4466 |
Silent Mutation |
AAC,AAT |
N1020N |
XP_016874600.1 |
XM_017019112.1 |
4466 |
Intron |
|
|
XP_016874601.1 |
View Full Product Details