Product Details
- SNP ID
-
rs2842030
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:163070705 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTACAAAAATACAAGAGTGTCAGGA[G/T]AAAGTTATGCTCTGGTAAATATTTT
- Phenotype
-
MIM: 602516
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RGS4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75931331] are located under a probe and SNP(s) [rs75708071] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RGS4
- Gene Name
- regulator of G-protein signaling 4
View Full Product Details