Product Details

SNP ID
rs10971520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33460848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAATTTATTCCCTCCCTATTCTAA[C/T]TTCAACCCTGGGCTGAGCTGCTCCA
Phenotype
MIM: 611532
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6851 PubMed Links

Gene Details

Gene
MIR6851
Gene Name
microRNA 6851
There are no transcripts associated with this gene.

Gene
NOL6
Gene Name
nucleolar protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022917.4 5314 Intron NP_075068.2
NM_139235.3 5314 Intron NP_631981.2
XM_017015044.1 5314 UTR 3 XP_016870533.1

View Full Product Details