Product Details

SNP ID
rs2458413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104347204 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGTGCCTTCATTTTAATTATTC[C/T]GATCTAGGAAAAAAGAAAAGATGAT
Phenotype
MIM: 605933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCSTAMP PubMed Links

Gene Details

Gene
DCSTAMP
Gene Name
dendrocyte expressed seven transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257317.1 Intron NP_001244246.1
NM_030788.3 Intron NP_110415.1
XM_011517321.1 Intron XP_011515623.1
XM_011517324.1 Intron XP_011515626.1
XM_017013878.1 Intron XP_016869367.1
XM_017013879.1 Intron XP_016869368.1
XM_017013880.1 Intron XP_016869369.1

View Full Product Details