Product Details
- SNP ID
-
rs34215158
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:102745962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACAAAATATAGAATATAGTAAAT[A/C]CAGAATTTACTTTTCCTTTGCACAT
- Phenotype
-
MIM: 615570
MIM: 611129
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SFXN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74557102] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SFXN2
- Gene Name
- sideroflexin 2
There are no transcripts associated with this gene.
- Gene
- WBP1L
- Gene Name
- WW domain binding protein 1-like
View Full Product Details