Product Details

SNP ID
rs2271625
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:85398390 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCCCGAACCCATTTGGAAATTAGG[A/T]GGATTCCAAATTACAAAGAAATGCA
Phenotype
MIM: 153615 MIM: 615427
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPG PubMed Links

Gene Details

Gene
CAPG
Gene Name
capping actin protein, gelsolin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256139.1 Intron NP_001243068.1
NM_001256140.1 Intron NP_001243069.1
NM_001320732.1 Intron NP_001307661.1
NM_001320733.1 Intron NP_001307662.1
NM_001320734.1 Intron NP_001307663.1
NM_001747.3 Intron NP_001738.2
XM_011533122.1 Intron XP_011531424.1
XM_011533123.1 Intron XP_011531425.1
Gene
ELMOD3
Gene Name
ELMO domain containing 3
There are no transcripts associated with this gene.

View Full Product Details