Product Details

SNP ID
rs10849268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5433141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAGCGCCCACCCGGTGGCCAC[C/G]CCACCCTGGGCCTTTGCGCAGATGT
Phenotype
MIM: 162660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NTF3 PubMed Links

Gene Details

Gene
NTF3
Gene Name
neurotrophin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102654.1 82 Intron NP_001096124.1
NM_002527.4 82 Intron NP_002518.1
XM_011520963.2 82 UTR 5 XP_011519265.1

View Full Product Details